Learn More
Medchemexpress LLC Paroxetine (hydrochloride) | 78246-49-8 | 100.0% | 1 ML
SDP
Catalog No. 5000406418
Encompass_Preferred
Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor and a GRK2 inhibitor with an IC50 of 14 μM. It is used in research for depressive disorders and other biological activities.
  • Potent selective serotonin-reuptake inhibitor
  • GRK2 inhibitor with an IC50 of 14 μM
  • Used for research of depressive disorder
  • Restrains T cell migration induced by CX3CL1
  • Inhibits GRK2 induced activation of ERK
  • Reduces pro-inflammatory cytokines in LPS-stimulated BV2 cells
  • Inhibits LPS-induced nitric oxide production and iNOS expression in BV2 cells
  • Blocks LPS-induced JNK activation
  • Attenuates symptoms of CIA rats and prevents histological damage of joints
  • Reduces myocyte cross-sectional area in rats and ROS formation
Catalog No. 50-004-06418 Supplier Medchemexpress LLC Supplier No. HYB049210MM/1ML
Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor and a GRK2 inhibitor with an IC50 of 14 μM. It is used in research for depressive disorders and other biological activities.
  • Potent selective serotonin-reuptake inhibitor
  • GRK2 inhibitor with an IC50 of 14 μM
  • Used for research of depressive disorder
  • Restrains T cell migration induced by CX3CL1
  • Inhibits GRK2 induced activation of ERK
  • Reduces pro-inflammatory cytokines in LPS-stimulated BV2 cells
  • Inhibits LPS-induced nitric oxide production and iNOS expression in BV2 cells
  • Blocks LPS-induced JNK activation
  • Attenuates symptoms of CIA rats and prevents histological damage of joints
  • Reduces myocyte cross-sectional area in rats and ROS formation