Learn More
Medchemexpress LLC Paroxetine (hydrochloride) | 78246-49-8 | 99.98% | 365.83 | 500 MG
SDP
Catalog No. 5000365916
Encompass_Preferred
Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor (SSRI), also known as a GRK2 inhibitor with an IC50 of 14 μM. It is commonly used for the research of depressive disorder.
  • Potent selective serotonin-reuptake inhibitor
  • GRK2 inhibitor with IC50 of 14 μM
  • Used for research of depressive disorder
  • Available in solid and solution forms
  • High purity of 99.98%
Catalog No. 50-003-65916 Supplier Medchemexpress LLC Supplier No. 500MGHYB0492
Paroxetine hydrochloride is a potent selective serotonin-reuptake inhibitor (SSRI), also known as a GRK2 inhibitor with an IC50 of 14 μM. It is commonly used for the research of depressive disorder.
  • Potent selective serotonin-reuptake inhibitor
  • GRK2 inhibitor with IC50 of 14 μM
  • Used for research of depressive disorder
  • Available in solid and solution forms
  • High purity of 99.98%