Learn More
Apexbio Technology LLC Paroxetine maleate 64006-44-6 50mg
SDP
Catalog No. 5000088844
Encompass_Preferred
Paroxetine maleate (CAS 64006-44-6) is a selective serotonin reuptake inhibitor (SSRI) that blocks the reabsorption of serotonin into presynaptic neurons resulting in elevated synaptic serotonin levels and modulation of neurotransmission This compound is widely utilized in the investigation of serotonergic signaling related to anxiety disorders including generalized anxiety disorder post-traumatic stress disorder and premenstrual dysphoric disorder Additionally paroxetine maleate serves as a research tool in studies probing the neuropharmacological mechanisms of chronic headache and its prospective therapeutic interventions
Catalog No. 50-000-88844 Supplier Apexbio Technology LLC Supplier No. B695250
Add to Cart
Add to Cart
Paroxetine maleate (CAS 64006-44-6) is a selective serotonin reuptake inhibitor (SSRI) that blocks the reabsorption of serotonin into presynaptic neurons resulting in elevated synaptic serotonin levels and modulation of neurotransmission This compound is widely utilized in the investigation of serotonergic signaling related to anxiety disorders including generalized anxiety disorder post-traumatic stress disorder and premenstrual dysphoric disorder Additionally paroxetine maleate serves as a research tool in studies probing the neuropharmacological mechanisms of chronic headache and its prospective therapeutic interventions