Learn More
Enzo Life Sciences PARP-1 (catalytic domain) (human), (recombinant) (His-tag) (20µg)
SDP
Catalog No. 502007952
Encompass_Preferred
Alternative name: Poly(ADP-ribose) polymerase 1. Purity: ≥90% (SDS-PAGE). MW: ~46kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 300mM sodium chloride, 250mM imidazole, 2mM β-mercaptoethanol, 0.2% NP-40 and 10% glycerol. Source: Produced in SF21 insect cells. Human PARP-1 (aa 656-1014) is fused to a C-terminal His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-7952 Supplier Enzo Life Sciences Supplier No. ALX201232C020
Alternative name: Poly(ADP-ribose) polymerase 1. Purity: ≥90% (SDS-PAGE). MW: ~46kDa. Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 300mM sodium chloride, 250mM imidazole, 2mM β-mercaptoethanol, 0.2% NP-40 and 10% glycerol. Source: Produced in SF21 insect cells. Human PARP-1 (aa 656-1014) is fused to a C-terminal His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.