Learn More
Enzo Life Sciences PARP-10 (human) monoclonal antibody (5H11) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX804626L001
Encompass_Preferred
Clone: 5H11. Isotype: IgG1. Immunogen: Recombinant human PARP-10 (poly(ADP-ribose) polymerase-10) (aa 1-907). Species reactivity: Human. Applications: ICC, IP, WB. Formulation: Liquid. Tissue culture supernatant (TCS) containing 0.1% sodium azide.
Catalog No. 50-200-9961