Learn More
Enzo Life Sciences PARP-1 polyclonal antibody (100 µl)
SDP
Supplier:  Enzo Life Sciences ALX210302R100
Encompass_Preferred
Host: Rabbit. Immunogen: Recombinant human PARP-1 (poly(ADP-ribose) polymerase-1) (aa 1-1014). UniProt: P09874. Species Reactivity: Human, Mouse, Monkey, Bovine. Crossreactivity: Does not cross-react with PARP-2. Applications: ELISA, ICC, IHC (FS), IHC (PS), IP, WB. Recommended dilutions: Immunocytochemistry (1:4,000). Immunoprecipitation (1:400). Western Blot (1:4,000). Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Formulation: Liquid. Neat serum containing 0.02% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. NC1469692