Learn More
AdipoGen PARP-2 ARTD2 ms pAb
SDP
Supplier:  AdipoGen AG25T0003R100
Encompass_Preferred
Polyclonal Antibody. Recognizes mouse PARP-2. Cross-reacts weakly with human PARP-2. Source Rabbit. Applications ICC, IHC, IP, WB. Liquid. Contains 0.1% sodium azide. PARP-2 ARTD2 is involved in the base excision repair BER pathway, by catalyzing the polyADP-ribosylation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks.
Catalog No. 50-256-3175