Learn More
Medchemexpress LLC Parp-2-in-1 | 2115698-83-2 | 99.6% | 465.40 g·mol⁻¹ | C21H19F4N5O3 | 10 MG
SDP
Catalog No. 5000247470
Encompass_Preferred
PARP-2-IN-1 is a potent, selective inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) used in research on DNA damage response and programmed cell death. It exhibits an IC50 of 11.5 nM against PARP-2 and is supplied as a high-purity small-molecule reagent for laboratory studies.
  • Potent and selective PARP-2 inhibition (IC50 11.5 nM).
  • High purity suitable for research (>99.5% by HPLC).
  • Well-characterized molecular properties (C21H19F4N5O3, 465.40 g·mol⁻¹).
  • Available in small mg-scale quantities for laboratory research.
  • Suitable for biochemical and cell-based assays studying DNA repair and apoptosis.
  • Provided with product datasheet and analytical data.
Catalog No. 50-002-47470 Supplier Medchemexpress LLC Supplier No. HY10203510MG
PARP-2-IN-1 is a potent, selective inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) used in research on DNA damage response and programmed cell death. It exhibits an IC50 of 11.5 nM against PARP-2 and is supplied as a high-purity small-molecule reagent for laboratory studies.
  • Potent and selective PARP-2 inhibition (IC50 11.5 nM).
  • High purity suitable for research (>99.5% by HPLC).
  • Well-characterized molecular properties (C21H19F4N5O3, 465.40 g·mol⁻¹).
  • Available in small mg-scale quantities for laboratory research.
  • Suitable for biochemical and cell-based assays studying DNA repair and apoptosis.
  • Provided with product datasheet and analytical data.