Learn More
Medchemexpress LLC Parp-2-in-1 | 2115698-83-2 | 99.6% | 465.40 g/mol | C21H19F4N5O3 | 5 MG
SDP
Catalog No. 5000247025
Encompass_Preferred
A potent, selective small-molecule inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) with a reported IC50 of 11.5 nM. The compound has molecular formula C21H19F4N5O3, molecular weight 465.40 g/mol, and is supplied as a white to off-white solid for use in biochemical and cellular research.
  • Potent PARP-2 inhibition (IC50 11.5 nM).
  • High chemical purity (99.6%).
  • Molecular formula C21H19F4N5O3 and molecular weight 465.40 g/mol for easy reference.
  • Supplied as a solid suitable for handling and storage.
  • Manufacturer documentation available: data sheet, COA, and SDS for characterization.
Catalog No. 50-002-47025 Supplier Medchemexpress LLC Supplier No. HY1020355MG
A potent, selective small-molecule inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) with a reported IC50 of 11.5 nM. The compound has molecular formula C21H19F4N5O3, molecular weight 465.40 g/mol, and is supplied as a white to off-white solid for use in biochemical and cellular research.
  • Potent PARP-2 inhibition (IC50 11.5 nM).
  • High chemical purity (99.6%).
  • Molecular formula C21H19F4N5O3 and molecular weight 465.40 g/mol for easy reference.
  • Supplied as a solid suitable for handling and storage.
  • Manufacturer documentation available: data sheet, COA, and SDS for characterization.