Learn More
Medchemexpress LLC Parp-2-in-1 | 2115698-83-2 | 99.6% | 465.40 g·mol⁻¹ | C21H19F4N5O3 | 50 MG
SDP
Catalog No. 5000247304
Encompass_Preferred
PARP-2-IN-1 is a potent, selective small-molecule inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) with a reported IC50 of 11.5 nM. Supplied as a high-purity powder, it is intended for biochemical and cellular research into DNA repair pathways and target validation studies.
  • Potent PARP-2 inhibition with low nanomolar IC50.
  • High chemical purity suitable for research use.
  • Available in multiple preweighed sizes for experimental flexibility.
  • Stable powder with defined cold-storage conditions.
  • Suitable for biochemical assays and cellular studies of DNA damage response.
Catalog No. 50-002-47304 Supplier Medchemexpress LLC Supplier No. HY10203550MG
PARP-2-IN-1 is a potent, selective small-molecule inhibitor of poly(ADP-ribose) polymerase 2 (PARP-2) with a reported IC50 of 11.5 nM. Supplied as a high-purity powder, it is intended for biochemical and cellular research into DNA repair pathways and target validation studies.
  • Potent PARP-2 inhibition with low nanomolar IC50.
  • High chemical purity suitable for research use.
  • Available in multiple preweighed sizes for experimental flexibility.
  • Stable powder with defined cold-storage conditions.
  • Suitable for biochemical assays and cellular studies of DNA damage response.