Learn More
Enzo Life Sciences PARP-2 monoclonal antibody (4G8) (1ml)
SDP
Supplier:  Enzo Life Sciences ALX804639L001
Encompass_Preferred
Clone: 4G8. Isotype: IgG1. Immunogen: Recombinant mouse PARP-2 (poly(ADP-ribose) polymerase-2). Species reactivity: Human, Mouse. Specificity: Recognizes an epitope in domain E of PARP-2. Applications: ELISA, WB. Formulation: Liquid. Tissue culture supernatant containing 0.1% sodium azide.
Catalog No. 50-200-9962