Learn More
Enzo Life Sciences PARP-2 (mouse), (recombinant) (high purity) (20µg)
SDP
Supplier:  Enzo Life Sciences ALX201064C020
Encompass_Preferred
Alternative name: Poly(ADP-ribose) polymerase 2. Concentration: ~1mg/ml. Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 1mM DTT, 0.5mM EDTA, 0.5mM PMSF and 20% glycerol. Source: Produced in Sf9 insect cells (full length). Produced in a baculovirus expression system. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7908