Learn More
AdipoGen PARP-2 ms Hishigh purity
SDP
Catalog No. 502562798
Encompass_Preferred
Protein. Mouse full-length PARP-2 ARTD2 is fused to a His-tag. Source Sf21 cells. Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride and 50mM imidazole, 0.2% NP-40 and 10% glycerol. Purity >98% SDS-PAGE. PARP-2 is involved in the base excision repair BER pathway, by catalyzing the polyADP-ribosylation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks.
Catalog No. 50-256-2798 Supplier AdipoGen Supplier No. AG40T0012C010
Protein. Mouse full-length PARP-2 ARTD2 is fused to a His-tag. Source Sf21 cells. Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride and 50mM imidazole, 0.2% NP-40 and 10% glycerol. Purity >98% SDS-PAGE. PARP-2 is involved in the base excision repair BER pathway, by catalyzing the polyADP-ribosylation of a limited number of acceptor proteins involved in chromatin architecture and in DNA metabolism. This modification follows DNA damages and appears as an obligatory step in a detection/signaling pathway leading to the reparation of DNA strand breaks.