Learn More
Enzo Life Sciences PARP-2 (mouse), (recombinant) (high purity) (His-tag) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX201251C010
Encompass_Preferred
Alternative name: Poly(ADP-ribose) polymerase 2. Concentration: 1mg/ml. Purity: ≥98% (SDS-PAGE). Formulation: Liquid. In 50mM TRIS-HCl, pH 7.5, containing 100mM sodium chloride, 50mM imidazole, 0.2% NP-40 and 10% glycerol. Source: Produced in Sf21 insect cells. Mouse full-length PARP-2 (aa 1-559) is fused to a C-terminus His-tag. Produced in a baculovirus expression system. Long Term Storage: -80°C. Handling: Avoid freeze/thaw cycles. After opening, prepare aliquots and store at -80°C.
Catalog No. 50-200-7958