Learn More
Genetex Inc PARP Monoclonal Antibody [HMV334] HistoMAX™ (GTX640566) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX640566
Encompass_Preferred
The PARP antibody [HMV334] HistoMAX™ (GTX640566) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human gastrointestinal stromal tumor (GIST) tissue using GTX640566 PARP antibody [HMV334] HistoMAX. Gastrointestinal stromal tumor (GIST) with strong PARP1 immunostaining of tumor cells.
Catalog No. NC3669363