Learn More
Santa Cruz Biotechnology PARP ANTIBODY MONOCLONAL
Supplier:  Santa Cruz Biotechnology SC8007
Encompass
Antibody; PARP-1 (F-2); Mouse monoclonal IgG2a;; Santa Cruz Biotech; 200ug/mL, Raised against amino acids 764-1014 mapping at the C-terminus of PARP-1 of human origin, fluorescein (sc-8007 FITC) and rhodamine (sc-8007 TRITC) conjugates
Catalog No. NC9295637