Learn More
Enzo Life Sciences PARP polyclonal antibody (25 µl)
SDP
Supplier:  Enzo Life Sciences BMLSA2530025
Encompass_Preferred
Immunogen: Synthetic peptide corresponding to aa 509-524 of bovine PARP automodification domain. Source: Whole rabbit serum. Species reactivity: Human, Mouse, Rat, Bovine. Applications: ELISA, WB. Formulation: Liquid. Neat serum containing 0.05% sodium azide.
Catalog No. 50-201-1780