Learn More
Sino Biological PARP-1 / PARP Antibody, Mouse MAb 100uL
Supplier:  Sino Biological 11040MM04100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PARP-1 / PARP (rh PARP-1 / PARP; Catalog#11040-H08B; NP_001609.2; Met 1-Trp 1014). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-224-8590