Learn More
Sino Biological PARP-1 / PARP Antibody, Rabbit PAb, Antigen Affinity Purified 100uL
Supplier:  Sino Biological 11040RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PARP1 (rh PARP1; Catalog#11040-H08B; NP_001609.2; Met 1-Trp 1014). PARP1 specific IgG was purified by human PARP1 affinity chromatography.
Catalog No. 50-224-8593