Learn More
Aviva Systems Biology Corp PARP10 Antibody - N-terminal region (ARP42811_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42811_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP10. It was validated on Western Blot. PARP10,poly (ADP-ribose) polymerase family, member 10,FLJ14464; PARP10,poly [ADP-ribose] polymerase 10,poly [ADP-ribose] polymerase 10,PARP-10The immunogen is a synthetic peptide directed towards the N terminal region of human PARP10. Alternate names: FLJ14464ApplicationWB. Species reactivity: Human, Rat
Catalog No. 50-285-028