Learn More
Aviva Systems Biology Corp PARP10 Antibody - middle region (ARP42810_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP42810_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP10. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PARP10. Alternate names: FLJ14464ApplicationWB. Species reactivity: Human
Catalog No. 50-014-58342