Learn More
TARGETMOL CHEMICALS INC PARP10-IN-2 5MG
SDP
Catalog No. 502579225
Encompass_Preferred
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PARP10-IN-2 is a potent inhibitor of PARP10 a mono-ADP-ribosyltransferase with an IC50 value of 3.64 uM for human PARP10.PARP10-IN-2 also inhibited PARP2 and PARP15 with IC50 values of 27 uM and 11 uM respectively. purity: 99%
Catalog No. 50-257-9225 Supplier TARGETMOL CHEMICALS INC Supplier No. T730075MG
Also available in 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. PARP10-IN-2 is a potent inhibitor of PARP10 a mono-ADP-ribosyltransferase with an IC50 value of 3.64 uM for human PARP10.PARP10-IN-2 also inhibited PARP2 and PARP15 with IC50 values of 27 uM and 11 uM respectively. purity: 99%