Learn More
Aviva Systems Biology Corp PARP11 Antibody - N-terminal region (ARP33768_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33768_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP11. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human PARP11. Alternate names: ARTD11, C12orf6ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55145