Learn More
Aviva Systems Biology Corp PARP12 Antibody - middle region (ARP33980_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33980_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP12. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PARP12. Alternate names: FLJ22693, MST109, MSTP109, PARP-12, ZC3H1, ZC3HDC1ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55213