Learn More
Medchemexpress LLC PARP14 inhibitor H10 | 2084811-68-5 | >98.0% | 557.58 | C24H27N7O7S | 5 MG
SDP
Catalog No. 5000226846
Encompass_Preferred
PARP14 inhibitor H10 is a selective small-molecule inhibitor of PARP14 used as a research tool to probe PARP14-dependent biology. It inhibits PARP14 with an IC50 of approximately 490 nM and has been reported to induce caspase-3/7-mediated apoptosis in cellular assays.
  • Selective PARP14 inhibition (IC50 ≈ 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated apoptosis in cellular assays.
  • High purity suitable for research (reported ≥98%).
  • Molecular formula C24H27N7O7S and molecular weight 557.58 g/mol.
Catalog No. 50-002-26846 Supplier Medchemexpress LLC Supplier No. HY1178895MG
PARP14 inhibitor H10 is a selective small-molecule inhibitor of PARP14 used as a research tool to probe PARP14-dependent biology. It inhibits PARP14 with an IC50 of approximately 490 nM and has been reported to induce caspase-3/7-mediated apoptosis in cellular assays.
  • Selective PARP14 inhibition (IC50 ≈ 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated apoptosis in cellular assays.
  • High purity suitable for research (reported ≥98%).
  • Molecular formula C24H27N7O7S and molecular weight 557.58 g/mol.