Learn More
Medchemexpress LLC Parp14 inhibitor H10 | 2084811-68-5 | 99.5% | 557.58 | C24H27N7O7S | 1 ML
SDP
Catalog No. 5000307970
Encompass_Preferred
PARP14 inhibitor H10 is a small-molecule inhibitor selective for poly(ADP-ribose) polymerase 14 (PARP14) that has demonstrated cellular activity and induction of caspase-3/7-mediated apoptosis. It is provided in ready-to-use and solid formats for laboratory research, with a high reported purity.
  • Selective PARP14 inhibition with reported IC50 around 490 nM.
  • Induces caspase-3/7-mediated apoptosis in cell-based assays.
  • Supplied as a 10 mM solution in DMSO for convenient in vitro use.
  • Available in multiple solid sizes for assay development and scale-up.
  • High reported purity supports reproducible experimental results.
Catalog No. 50-003-07970 Supplier Medchemexpress LLC Supplier No. HY11788910MM 1ML
PARP14 inhibitor H10 is a small-molecule inhibitor selective for poly(ADP-ribose) polymerase 14 (PARP14) that has demonstrated cellular activity and induction of caspase-3/7-mediated apoptosis. It is provided in ready-to-use and solid formats for laboratory research, with a high reported purity.
  • Selective PARP14 inhibition with reported IC50 around 490 nM.
  • Induces caspase-3/7-mediated apoptosis in cell-based assays.
  • Supplied as a 10 mM solution in DMSO for convenient in vitro use.
  • Available in multiple solid sizes for assay development and scale-up.
  • High reported purity supports reproducible experimental results.