Learn More
Medchemexpress LLC PARP14 inhibitor H10 | 2084811-68-5 | 99.5% | 557.58 | C24H27N7O7S | 10 MG
SDP
Catalog No. 5000227014
Encompass_Preferred
A selective small-molecule inhibitor of PARP14 designed for biochemical and cellular research targeting PARP14-mediated pathways and apoptosis.
  • Selective PARP14 inhibitor with reported IC50 of 490 nM.
  • High reported purity (99.46% as supplied).
  • Molecular weight 557.58 and formula C24H27N7O7S.
  • Supplied as a solid (off-white to yellow) or as a 10 mM solution in DMSO for easy reconstitution.
  • Available in small research quantities, including 10 mg.
Catalog No. 50-002-27014 Supplier Medchemexpress LLC Supplier No. HY11788910MG
A selective small-molecule inhibitor of PARP14 designed for biochemical and cellular research targeting PARP14-mediated pathways and apoptosis.
  • Selective PARP14 inhibitor with reported IC50 of 490 nM.
  • High reported purity (99.46% as supplied).
  • Molecular weight 557.58 and formula C24H27N7O7S.
  • Supplied as a solid (off-white to yellow) or as a 10 mM solution in DMSO for easy reconstitution.
  • Available in small research quantities, including 10 mg.