Learn More
Medchemexpress LLC PARP14 inhibitor H10 | 2084811-68-5 | 99.5% | 557.58 | C24H27N7O7S | 50 MG
SDP
Catalog No. 5000227653
Encompass_Preferred
PARP14 inhibitor H10 is a selective small-molecule research inhibitor of Poly(ADP-ribose) polymerase 14 (PARP14). It inhibits PARP14 with an IC50 of 490 nM and shows approximately 24-fold selectivity over PARP1. The compound is provided as a high-purity solid and is soluble in dimethyl sulfoxide (DMSO).
  • Selective inhibition of PARP14 with IC50 = 490 nM.
  • Approximately 24-fold selectivity over PARP1.
  • High purity (≈99.5%).
  • Soluble in DMSO at about 50 mg/mL; ultrasonic assistance recommended.
  • Available in small research quantities, including 50 mg.
Catalog No. 50-002-27653 Supplier Medchemexpress LLC Supplier No. HY11788950MG
Add to Cart
Add to Cart
PARP14 inhibitor H10 is a selective small-molecule research inhibitor of Poly(ADP-ribose) polymerase 14 (PARP14). It inhibits PARP14 with an IC50 of 490 nM and shows approximately 24-fold selectivity over PARP1. The compound is provided as a high-purity solid and is soluble in dimethyl sulfoxide (DMSO).
  • Selective inhibition of PARP14 with IC50 = 490 nM.
  • Approximately 24-fold selectivity over PARP1.
  • High purity (≈99.5%).
  • Soluble in DMSO at about 50 mg/mL; ultrasonic assistance recommended.
  • Available in small research quantities, including 50 mg.