Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PARP14 inhibitor H10 | 2084811-68-5 | 99.5% | 557.58 g/mol | C24H27N7O7S | 100 MG
SDP

Catalog No. 5000227562
Encompass_Preferred

PARP14 inhibitor H10 is a selective small-molecule research compound that targets PARP14 and induces caspase-3/7-mediated apoptosis in cellular assays. It is supplied as a solid and is intended for laboratory research use only.

  • Selective PARP14 inhibition with reported IC50 = 490 nM.
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated apoptosis in cell-based assays.
  • High reported purity suitable for research (≈99.5%).
  • Molecular weight 557.58 g/mol; formula C24H27N7O7S.
  • Soluble in DMSO; supplied in sizes up to 100 mg for experimental use.

Catalog No. 50-002-27562 Supplier Medchemexpress LLC Supplier No. HY117889100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Request a Quote

PARP14 inhibitor H10 is a selective small-molecule research compound that targets PARP14 and induces caspase-3/7-mediated apoptosis in cellular assays. It is supplied as a solid and is intended for laboratory research use only.

  • Selective PARP14 inhibition with reported IC50 = 490 nM.
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated apoptosis in cell-based assays.
  • High reported purity suitable for research (≈99.5%).
  • Molecular weight 557.58 g/mol; formula C24H27N7O7S.
  • Soluble in DMSO; supplied in sizes up to 100 mg for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.