Learn More
Medchemexpress LLC PARP14 inhibitor H10 | 2084811-68-5 | 99.5% | 557.58 g/mol | C24H27N7O7S | 25 MG
SDP
Catalog No. 5000228138
Encompass_Preferred
PARP14 inhibitor H10 is a research-grade small-molecule inhibitor selective for PARP14 (IC50 ≈ 490 nM) that demonstrates approximately 24-fold selectivity over PARP1. Supplied as a solid for laboratory research use, the compound is provided with analytical documentation and safety data for handling.
  • Selective PARP14 inhibition (IC50 ≈ 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated cell apoptosis.
  • High purity (~99.5%) with HNMR, LCMS, and RP-HPLC characterization.
  • Available in mg pack sizes for research use only.
  • Includes datasheet, certificate of analysis, and SDS for handling and safety.
Catalog No. 50-002-28138 Supplier Medchemexpress LLC Supplier No. HY11788925MG
PARP14 inhibitor H10 is a research-grade small-molecule inhibitor selective for PARP14 (IC50 ≈ 490 nM) that demonstrates approximately 24-fold selectivity over PARP1. Supplied as a solid for laboratory research use, the compound is provided with analytical documentation and safety data for handling.
  • Selective PARP14 inhibition (IC50 ≈ 490 nM).
  • Approximately 24-fold selectivity over PARP1.
  • Induces caspase-3/7-mediated cell apoptosis.
  • High purity (~99.5%) with HNMR, LCMS, and RP-HPLC characterization.
  • Available in mg pack sizes for research use only.
  • Includes datasheet, certificate of analysis, and SDS for handling and safety.