Learn More
Aviva Systems Biology Corp PARP16 Antibody - middle region (ARP33752_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33752_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP16. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PARP16. Alternate names: C15orf30, FLJ20509, FLJ25281, pART15ApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55137