Learn More
Cayman Chemical PARP1humnrecombInt 20ug
SDP
Catalog No. 502946238
Encompass_Preferred
Active recombinant human N-terminal GST-tagged PARP1 expressed in insect cells • Amino acids: 2-1,041 (full length) • MW: 139 kDa
Catalog No. 50-294-6238 Supplier Cayman Chemical Supplier No. 3256120 UG
Active recombinant human N-terminal GST-tagged PARP1 expressed in insect cells • Amino acids: 2-1,041 (full length) • MW: 139 kDa