Learn More
Aviva Systems Biology Corp PARP2 Antibody - middle region (ARP33758_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33758_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human PARP2. Alternate names: ADPRT2, PARP-2, ADPRTL2, ADPRTL3, pADPRT-2ApplicationIF, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52925