Learn More
Aviva Systems Biology Corp PARP3 Antibody - C-terminal region (ARP33760_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33760_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PARP3. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human PARP3. Alternate names: IRT1, ADPRT3, ADPRTL2, ADPRTL3, PADPRT-3ApplicationIHC, WB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-52926