Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Protein mono-ADP-ribosyltransferase TIPARP (PARP7) | 10UG
SDP

Catalog No. 5000175101
Encompass_Preferred

Recombinant human PARP7 protein fragment with His and Avi affinity tags, expressed in E. coli. The construct covers residues M456-I657 (UniProt Q7Z3E1) and is supplied in a stabilized, filtered buffer suitable for biochemical assays, binding studies, and structural work.

  • Contains residues M456-I657 of human PARP7 (UniProt Q7Z3E1).
  • Expressed in E. coli for efficient recombinant production.
  • Includes His and Avi tags for affinity purification and detection.
  • Supplied in 50 mM HEPES pH 7.5, 500 mM NaCl, 1 mM DTT, 20% glycerol; 0.22 μm filtered.
  • Store at -80 °C for long-term stability; stable at -20 °C for ~3 months after opening.
  • Approximate molecular weight 28.3 kDa.

Catalog No. 50-001-75101 Supplier Medchemexpress LLC Supplier No. HYP70362210UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant human PARP7 protein fragment with His and Avi affinity tags, expressed in E. coli. The construct covers residues M456-I657 (UniProt Q7Z3E1) and is supplied in a stabilized, filtered buffer suitable for biochemical assays, binding studies, and structural work.

  • Contains residues M456-I657 of human PARP7 (UniProt Q7Z3E1).
  • Expressed in E. coli for efficient recombinant production.
  • Includes His and Avi tags for affinity purification and detection.
  • Supplied in 50 mM HEPES pH 7.5, 500 mM NaCl, 1 mM DTT, 20% glycerol; 0.22 μm filtered.
  • Store at -80 °C for long-term stability; stable at -20 °C for ~3 months after opening.
  • Approximate molecular weight 28.3 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.