Learn More
Medchemexpress LLC HY-109068 10mg Medchemexpress, Parsaclisib CAS:1426698-88-5 Purity:>98%
SDP
Catalog No. 501968664
Encompass_Preferred
Medchemexpress, HY-109068 10mg Parsaclisib CAS:1426698-88-5 Parsaclisib is a potent and selective PI3Kδ inhibitor, with an IC50 of 1 nM at 1 mM ATP, and shows appr 20,000-fold selectivity for PI3Kα, PI3Kβ, PI3Kγ and 57 other kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8664 Supplier Medchemexpress LLC Supplier No. HY10906810MG
Medchemexpress, HY-109068 10mg Parsaclisib CAS:1426698-88-5 Parsaclisib is a potent and selective PI3Kδ inhibitor, with an IC50 of 1 nM at 1 mM ATP, and shows appr 20,000-fold selectivity for PI3Kα, PI3Kβ, PI3Kγ and 57 other kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.