Learn More
Medchemexpress LLC Parsaclisib hydrochloride | 1995889-48-9 | 99.3% | 469.34 | 1 MG
SDP
Catalog No. 5000343309
Encompass_Preferred
Parsaclisib hydrochloride is a potent, selective, and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. It exhibits approximately 20000-fold selectivity over other PI3K class I isoforms and is used for the research of relapsed or refractory B-cell malignancies.
  • Potent, selective, and orally active inhibitor of PI3Kδ
  • High potency and selectivity for PI3Kδ
  • Used for research of relapsed or refractory B-cell malignancies
  • Inhibits proliferation of MCL and DLBCL cell lines
  • Inhibits tumor growth in BALB/c mice bearing A20 murine lymphoma cells
  • Potential therapeutic agent for B-cell malignancies
Catalog No. 50-003-43309 Supplier Medchemexpress LLC Supplier No. HY109068A10MG
Parsaclisib hydrochloride is a potent, selective, and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. It exhibits approximately 20000-fold selectivity over other PI3K class I isoforms and is used for the research of relapsed or refractory B-cell malignancies.
  • Potent, selective, and orally active inhibitor of PI3Kδ
  • High potency and selectivity for PI3Kδ
  • Used for research of relapsed or refractory B-cell malignancies
  • Inhibits proliferation of MCL and DLBCL cell lines
  • Inhibits tumor growth in BALB/c mice bearing A20 murine lymphoma cells
  • Potential therapeutic agent for B-cell malignancies