Learn More
Medchemexpress LLC Parsaclisib hydrochloride | 1995889-48-9 | 99.3% | 469.34 | 1 ML
SDP
Catalog No. 5000343827
Encompass_Preferred
Parsaclisib hydrochloride (INCB050465 hydrochloride) is a potent, selective, and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. It exhibits approximately 20000-fold selectivity over other PI3K class I isoforms. It is used for research on relapsed or refractory B-cell malignancies.
  • Potent and selective PI3Kδ inhibitor
  • Exhibits high selectivity over other PI3K class I isoforms
  • Suitable for research on B-cell malignancies
  • Available for in vitro and in vivo studies
Catalog No. 50-003-43827 Supplier Medchemexpress LLC Supplier No. HY109068A100MG
Parsaclisib hydrochloride (INCB050465 hydrochloride) is a potent, selective, and orally active inhibitor of PI3Kδ, with an IC50 of 1 nM at 1 mM ATP. It exhibits approximately 20000-fold selectivity over other PI3K class I isoforms. It is used for research on relapsed or refractory B-cell malignancies.
  • Potent and selective PI3Kδ inhibitor
  • Exhibits high selectivity over other PI3K class I isoforms
  • Suitable for research on B-cell malignancies
  • Available for in vitro and in vivo studies