Learn More
Nova Biomedical Corporation Nova Biomedical Part control level 4
Catalog No. NC9816269
Encompass
Nova Biomedical - 24617 - BioProfile Control Level 4 30 ampules/box (contains Glutamine, Glutamate, Glucose, Lactate)
Catalog No. NC9816269 Supplier Nova Biomedical Corporation Supplier No. 24617
Nova Biomedical - 24617 - BioProfile Control Level 4 30 ampules/box (contains Glutamine, Glutamate, Glucose, Lactate)