Learn More
Cayman Chemical Parthenolide, Purity: ≥98%, CAS: 20554-84-1, 100 mg
SDP
Catalog No. 502896712
Encompass_Preferred
A natural sesquiterpene lactone which inhibits the growth of the promastigote form of L. amazonensis (IC50 = 3.6 μg/ml); induces apoptosis in cancer cells, at least in part by inhibiting NF-κB- and STAT-mediated anti-apoptotic gene transcription; directly binds the pattern recognition receptor NOD2
Catalog No. 50-289-6712 Supplier Cayman Chemical Supplier No. 70080100 MG
A natural sesquiterpene lactone which inhibits the growth of the promastigote form of L. amazonensis (IC50 = 3.6 μg/ml); induces apoptosis in cancer cells, at least in part by inhibiting NF-κB- and STAT-mediated anti-apoptotic gene transcription; directly binds the pattern recognition receptor NOD2