Learn More
AdipoGen Parthenolide,Chemical. CAS: 20554-84-1
SDP
Supplier:  AdipoGen AGCN20455M010
Encompass_Preferred
Parthenolide, AdipoGen Life Sciences. Chemical. CAS: 20554-84-1. Formula: C15H20O3. MW: 248.3. Isolated from Magnolia grandiflora. Anticancer and antiangiogenic compound. Induces apoptosis, cell cycle arrest and autophagy in various cancer cell lines. At low concentrations acts as antioxidant reducing oxidative stress generated through the TCR signaling pathway. At high concentration induces O2- and causes oxidative-stress-mediated apoptosis. Potent anti-inflammatory and anti-atherosclerotic agent. NF-kappaB inhibitor by directly targeting IkappaB kinase. Inhibits STAT-mediated anti-apoptotic gene transcription. NLRP3 inflammasome inhibitor. Inhibits ATPase activity of NLRP3 and protease activity of caspase 1. Directly binds the pattern recognition receptor NOD2. Specifically inhibits HDAC1 without affecting other class I/II HDACs. DNA methyltransferase 1 (DNMT1) inhibitor.
Catalog No. 50-114-6344