Learn More
Apexbio Technology LLC Parthenolide 20554-84-1 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC N13155.1
Encompass_Preferred
Parthenolide (CAS 20554-84-1) is a sesquiterpene lactone naturally derived from feverfew (Chrysanthemum parthenium) It exerts biological activity primarily via inhibition of the NF- B signaling pathway suppression of STAT3 activation and modulation of oxidative stress pathways leading to anti-inflammatory and anticancer effects In biomedical research parthenolide is commonly utilized to investigate molecular mechanisms governing inflammation cellular apoptosis and tumor growth suppression as well as in studies targeting leukemia and solid tumors
Catalog No. 50-000-63288