Learn More
TARGETMOL CHEMICALS INC Parthenolide 200MG
SDP
Catalog No. 502879614
Encompass_Preferred
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Parthenolide ((-)-Parthenolide) is a natural sesquiterpene lactone that is an NF-kB inhibitor and also specifically inhibits HDAC1 protein, but is inactive against other class I/II HDACs. Parthenolide has anti-inflammatory, anti-tumor, and anti-viral activities. Purity 99.33%
Catalog No. 50-287-9614 Supplier TARGETMOL CHEMICALS INC Supplier No. T2140200MG
Also available in 1 mL, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Parthenolide ((-)-Parthenolide) is a natural sesquiterpene lactone that is an NF-kB inhibitor and also specifically inhibits HDAC1 protein, but is inactive against other class I/II HDACs. Parthenolide has anti-inflammatory, anti-tumor, and anti-viral activities. Purity 99.33%