Learn More
Medchemexpress LLC HY-N0141 50mg Medchemexpress, Parthenolide CAS:20554-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN014150MG
Encompass_Preferred
Medchemexpress, HY-N0141 50mg Parthenolide CAS:20554-84-1 Parthenolide is a sesquiterpene lactone found in the medicinal herb Feverfew. Parthenolide exhibits anti-inflammatory activity by inhibiting NF-κB activation; also inhibits HDAC1 protein without affecting other class I/II HDACs. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1309