Learn More
Selleck Chemical LLC (-)-Parthenolide 100mg 20554-84-1
SDP
Supplier:  Selleck Chemical LLC S2341100MG
Encompass_Preferred
(-)-Parthenolide, an inhibitor of the Nuclear Factor-?B Pathway, specifically depletes HDAC1 protein without affecting other class I/II HDACs; Also promotes the ubiquitination of MDM2 and activates p53 cellular functions. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-530-2