Learn More
Thermo Scientific™ Particle-Certified Glass Containers

Description
Protect the quality of your products or lab samples. Thermo Scientific™ Particle-Certified Glass Containers are tested and certified to meet the low particle criteria; assembled to contain as few as 5 particles per milliliter > 0.5 microns. A large variant of standard particle-certified containers are available. Non-standard containers may be custom cleaned on request.
Key Features:
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Assembled with low-shedding polypropylene septa with chemically inert PTFE faced liners that do not contain adhesives
- Individual bottles double bagged to maintain integrity until point of use
- 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
Recommended for:
- Low particle applications
Specifications
Specifications
| Capacity | 32 oz., 1000 mL |
| Capacity (Metric) | 1000 mL |
| Capacity (English) | 32 oz. |
| Color | Clear |
| Description | Clear Glass Wide Mouth Jar with Polypropylene Closure, 1000 mL |
| Diameter (English) | 3.75 in. |
| Material | Glass |
| No. per Case | 12 |
| Mouth | Wide |
| Height (Metric) | 168.28 mm |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.