Learn More
PCE AMERICAS INC Air Sampler Measuring range 0 particles/28.3 l ... 1000000 particles/28.3 litres
SDP
Catalog No. NC3654591
Encompass_Preferred
"The state-of-the-art particle counter enables precise real-time measurement in the range of 0.3 μm to 10 μm to provide a comprehensive picture of air quality. The results are presented in two units, either number per 28.3 litres or number per cubic metre. Air sampling from the particle counter is done by an internal pump.Real-time measurement from 0.3 μm … 10 μmOutput in particles/28.3L or particles/m³Visual and audible alarm when threshold value is exceededModbus RTU and MQTT via RS485Temperature and humidity measurement"
Catalog No. NC3654591 Supplier PCE AMERICAS INC Supplier No. PCECPC 100
Add to Cart
Add to Cart
Request a Quote
"The state-of-the-art particle counter enables precise real-time measurement in the range of 0.3 μm to 10 μm to provide a comprehensive picture of air quality. The results are presented in two units, either number per 28.3 litres or number per cubic metre. Air sampling from the particle counter is done by an internal pump.Real-time measurement from 0.3 μm … 10 μmOutput in particles/28.3L or particles/m³Visual and audible alarm when threshold value is exceededModbus RTU and MQTT via RS485Temperature and humidity measurement"