Learn More
PCE AMERICAS INC Air Sampler 0.3 µm / 0.5 µm / 1.0 µm / 2.5 µm / 5.0 µm / 10 µm
SDP
Supplier:  PCE AMERICAS INC PCEPCO 1
Encompass_Preferred
"This is a laser particle counter, air sampler or dust measuring device for measuring the concentration of particles in the air. The dust measuring device is ideal for monitoring clean rooms, indoor air quality, exposure to exhaust, tobacco or cigarette smoke and other harmful air pollutants, and levels of airborne dust. Cigarette smoke particles mainly are distributed in the size range from 0.1 to 1.0 micron.A variety of materials are explosible in dust form. Some examples include: food (e.g., candy, sugar, spice, starch, flour and feed), grain, tobacco, plastics, wood, paper, pulp, rubber, pesticides, pharmaceuticals, dyes, coal and metals (e.g., aluminum, chromium, iron, magnesium and zinc). These materials are used in agriculture, chemical manufacturing, pharmaceutical production, furniture, textiles, fossil fuel power generation, recycling operations, metal working and processing (including additive manufacturing and 3D printing)
Catalog No. NC3654426