Learn More
PCE AMERICAS INC Air Sampler 0.3 µm / 0.5 µm / 1.0 µm / 2.5 µm / 5.0 µm / 10 µm | PM Channels PM 2.5, PM 10
SDP
Catalog No. NC3654439
Encompass_Preferred
"Portable handheld device for monitoring particulate matter (PM) concentrations in the air / 0.3 µm, 0.5 µm, 1.0 µm, 2.5 µm, 5.0 µm and 10 µm particle sizes / Measures air temperature, dew point and relative humidityThis is a portable handheld air sampler or particle counter used to monitor particulate matter (PM) concentrations in the air. Designed to aid in indoor air quality (IAQ) assessments, this particle counter also measures air temperature, dew point and relative humidity.The particle counter is ideal for monitoring clean rooms, exposure to exhaust, smoke and other harmful air pollutants, and levels of airborne combustible dust such as agricultural dust, carbonaceous dust, chemical dust, metal dust, plastic dust and wood dust."
Catalog No. NC3654439 Supplier PCE AMERICAS INC Supplier No. PCEPCO 2
"Portable handheld device for monitoring particulate matter (PM) concentrations in the air / 0.3 µm, 0.5 µm, 1.0 µm, 2.5 µm, 5.0 µm and 10 µm particle sizes / Measures air temperature, dew point and relative humidityThis is a portable handheld air sampler or particle counter used to monitor particulate matter (PM) concentrations in the air. Designed to aid in indoor air quality (IAQ) assessments, this particle counter also measures air temperature, dew point and relative humidity.The particle counter is ideal for monitoring clean rooms, exposure to exhaust, smoke and other harmful air pollutants, and levels of airborne combustible dust such as agricultural dust, carbonaceous dust, chemical dust, metal dust, plastic dust and wood dust."