Learn More
Cole-Parmer Digi-Sense Mini Particle Counter with Bluetooth Connectivity
Catalog No. 501961395 Shop All Cole-Parmer Products
Encompass_Preferred
Using our free Digi-Sense D/S Particle app, this mini particle counter with Bluetooth technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your workideal for record keeping at large sites.
Catalog No. 50-196-1395 Supplier Cole-Parmer Supplier No. 2025065
Using our free Digi-Sense D/S Particle app, this mini particle counter with Bluetooth technology allows you to transmit data directly to your Android or iOS device. Now you can view data at a safe distance from dangerous equipment or hazardous locations and even use your smart device as a real-time secondary display for checking measurements, eliminating the need to write down readings on paper. The results are automatically stored in the app with a date-and-time stamp and can be saved as a CSV file and emailed for future reference, manipulation, or analysis to help determine trends and conditions over a long period of time. Use the app to attach photos and notes to the records creating a clear reference point for your workideal for record keeping at large sites.